Learn More
Thermo Scientific Chemicals Cefotaxime sodium salt, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• The cefotaxime sodium salt is a - lactamase considered a third-generation cephalosporin with antibiotic and bactericidal activities
• This compound exerts its biological activity by binding to penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall leading to their inactivation. The inactivation of these proteins affects the cross-linking of peptidoglycan chains required for bacterial cell wall strength and rigidity, resulting in the weakening of cell walls and cell lysis.
Applications
• It is an efficient antibacterial compound that acts against gram-negative bacteria, without activity against gram-positive bacteria such as pseudomonas and penicillin-resistant strains of streptococcus pneumonia
• It has been used for plant cell culture and human cell lines in vitro
• Cefotaxime sodium salt has been used in antibiotic susceptibility testing (AST) methods
• Cefotaxime sodium salt is also used to remove bacteria from Ostreococcus tauri cultures
Specifications
Specifications
Type | Cefotaxime Sodium Salt |
Melting Point | 201.0°C to 210.0°C |
Packaging | Glass Bottle |
Quantity | 25 g |
Assay Percent Range | 95% |
Solubility Information | Solubility in water: soluble. |
IUPAC Name | sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Formula Weight | 477.44 |
Percent Purity | 95% |
Chemical Name or Material | Cefotaxime sodium salt |
Safety and Handling
- Cefotaxime sodium salt
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.