missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Celastrol, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Antioxidant and anti-inflammatory agent. Potently inhibits lipid peroxidation in mitochondria and inhibits TNF-α-induced NFκB activation. Also shown to inhibit topoisomerase II activity in vitro (IC50 = 7.41 μM).
Specifications
Quantity | 10 mg |
Formula Weight | 450.61 |
Percent Purity | >99% |
Chemical Name or Material | Celastrol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'