missing translation for 'onlineSavingsMsg'
Learn More

Celestine Blue, pure, Thermo Scientific Chemicals

Product Code. 10424795
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1562-90-9
Molecular Formula C17H18ClN3O4
Molecular Weight (g/mol) 363.798
MDL Number MFCD00011927
InChI Key OJAMERVLRXYMPD-UHFFFAOYSA-N
PubChem CID 54684697
IUPAC Name 7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride
SMILES CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-]

Specifications

Merck Index 15, 1961
Infrared Spectrum Authentic
Lambda Max 638 to 648nm
Formula Weight 363.79
Grade Extra Pure
Packaging Glass bottle
Specific Extinction 700 min. (in water)
UV Spectrometry Complies
Color Black to Brown
Quantity 50 g
Physical Form Powder
Chemical Name or Material Celestine blue
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'