Learn More
Velp Scientifica™ Celite™ 545
Velp Scientifica™ Celite™ 545 for use with HU 6 Hydrolysis Unit.
Brand: Velp Scientifica™
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Velp Scientifica™ Celite™ 545 for use with HU 6 Hydrolysis Unit.
- For use with HU 6 Hydrolysis Unit
Specifications
Celite 545 | |
1 kg | |
1 kg |
HU6 Hydrolysis Unit | |
1 Ea. |
Safety and Handling
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P314-Get medical advice/attention if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : diatomaceous earth (calcined), christobalit, quartz