missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Celite™ 545 Filter Aid, Flux-Calcined, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Quantity | 1 kg |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Diatomaceous Earth, Flux-Calcined, Treated with sodium carbonate filter aid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'