missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cell Proliferation Kit, BioTrak™ ELISA Kit and Components

Provides a precise, fast and simple colorimetric assay to quantitate proliferating cells cultured in a 96 well microplate

Brand:  Cytiva RPN201

173.85 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10218714

  • £192.00 / 10mL
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: The cell proliferation kit includes:
Labeling reagent, Nuclease, Anti-5-bromo-2'-deoxyuridine monoclonal antibody, Peroxidase anti-mouse IgG2a, Peroxidase anti-mouse IgG2a diluent, Substrate/intensifier

Cel proliferation Biotrak ELISA kit includes:
BrdU labelling reagent; Fixative; Blocking reagent; Peroxidase-labelled anti-BrdU; Antibody dilution solution; Wash buffer; Substrate solution

Cell proliferation labelling reagent and Anti-BrdU antibody are available separately
This item is not returnable. View return policy

Description

Description

  • Recommended cell number: 10e6-10e7 cells/mL (Cell proliferation kit) 10e2 cells/well-10e5 cells/well (Biotrak ELISA kit)
  • Required time: 2-3 hour post-labeling (labeling 1-2 hours)
  • Antibody and labeling reagent available separately for ELISA and ICC systems
  • Suitable for in vivo and in vitro tissue labeling in addition to cultured cells (ELISA format)
  • Non radioactive protocols provide sensitivity comparable to autoradiographic methods
  • Cell proliferation kit avoids the need to chemically denature DNA. Cellular morphology is preserved allowing multiple labeling experiments
  • 5-fluoro-2'-deoxyuridine is used to inhibit thymidilate synthetase increasing BrdU incorporation

When ordering the Cell Proliferation Kit (RPN20), you must also order DAB REAGENT OF RPN20/RPN1174 (RPNK8888)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10mL of Aqueous 5-bromo-2'-deoxyuridine and 5-fluoro-2'-deoxyuridine
10 mL
2°C to 8°C
Immunocytochemistry
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.