missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Recovery Solution
GREENER_CHOICE

Allows for the recovery of cells cultured on Corning™ Matrigel™ Basement Membrane Matrix for subsequent biochemical analyses

Brand:  Corning™ 354253

 View more versions of this product

Product Code. 11543560

  • £95.63 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
Cell recovery solution
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Cell Recovery Solution > Cell Recovery Solution; 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.