missing translation for 'onlineSavingsMsg'
Learn More

Hellma Cell for Scattered Light Measurements, Type 40-QS

Highly pure Suprasil™ quartz glass scattered light cell with a PTFE stopper.

Brand:  Hellma 540-110-80

Additional Details : Weight : 0.25000kg

Product Code. 10066001

  • £151.00 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

  • High-purity Suprasil™ quartz construction
  • UV/VIS range
  • For scattered light assays
  • PTFE stopper included
Specifications

Specifications

3200 μL
PTFE
Base Thickness: 1 mm
1/Pk
75 mm
Suprasil Quartz
8 mm
PTFE Stopper
200 - 2500 nm
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers