Learn More
Cell Viability Incorporated™ CCD Cell Viability Reagent Pack
Save money without losing performance with alternative reagents for automated CCD cell viability analyzers. Cell Viability Incorporated™ CCD Cell Viability Reagent Pack features long-term material stability.
Brand:  Cell Viability Incorporated™ 2400102
Code : LE
Additional Details : CAS Number : 67-63-0 Weight : 5.96250kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Product Code. 12740616
  • £340.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • Same count performance as original reagent pack vendor.
  • Suitable for Vi-Cell™ analyzers.
  • Long-term material stability - no cooling necessary.
  • Cost savings.
Specifications
Specifications
Designed as a direct replacement for the Vi-Cell™ Analyzer reagent pack.
Includes reagent kit and 200 sample cups.
Cell Viability Reagent Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Cell Viability Reagent Pack
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
Supplemental information
  • MIXTURE LIST-Contains : isopropyl alcohol