missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ HYPERStack™ Cell Culture Vessels - New Design
GREENER_CHOICEBIOPHARMA

More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint.

Brand:  Corning™ 20036

 View more versions of this product

Product Code. 17213084

  • £3100.00 / Pack of 2
Estimated Shipment: 14-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning's High Yield PERformance (HYPER) Platform - Closed System for High Yield Cell Growth

The Corning HYPERStack cell culture vessel combines the best of two Corning products: the Corning CellSTACK™ and the Corning HYPERFlask™ cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be among the most efficient, scalable cell culture vessels for adherent cell culture available today.

Features and Benefits

  • More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings support scale-up and scale-out
  • Ergonomic design – easier manipulation with handling equipment and accessories
  • Fixed media volume – 0.2 mL/cm2 fills vessel for less volumetric waste

Surface Treatment Guide

Corning CellBIND™ surface treatment is designed to enhance cell attachment and increase cell yields, improving attachment of loosely adherent cells and maintaining growth in reduced-serum or serum-free conditions. Alternatively, a non-treated option can support conventional cell attachment and allow for a faster and gentler harvest procedure for downstream processing and assays.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3920 mL
2/Cs.
36-layer vessel
Sterile
CellBIND
Polystyrene
1 Ea.
Rectangular
18,000 cm2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2213035 Certificate TypeCertificate of Quality Date02/11/2024 Catalog Number20036
Lot Number3324007 Certificate TypeCertificate of Quality Date29/05/2024 Catalog Number20036
Lot Number4091020 Certificate TypeCertificate of Compliance Date17/07/2024 Catalog Number20036
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CellBIND™ HYPERStack™ Cell Culture Vessels - New Design > Cell Culture Vessel; Layers: 36; Surface Treatment: CellBIND; Design: New

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.