missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
5/Pk., 20/Cs.
Description
- Improves adhesion of different cell types to polystyrene surfaces
- Reduced-serum or serum-free cultivation
- Increase in isolation and cultivation efficiency
- Ready-to-use products, time-saving
- Consistent quality
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Format | Standard |
Lid | With Lid |
Array | 16 x 24 |
Autoclavable | Not Autoclavable |
Packaging | 5/Pk., 20/Cs. |
Color | Clear |
Material | Polystyrene |
No. of Wells | 384 |
Volume (Metric) Working | 15 to 110 μL |
Sterility | Non-sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'