missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ 96-well, Poly-L Lysine-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Brand: Greiner Bio-One 655936
Additional Details : Weight : 1.08900kg
Description
Poly-L-Lysine (PLL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.- Coated with Poly-L-Lysine synthetic protein (30-70 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Standard | |
Room Temperature | |
8 x 12 | |
Polystyrene | |
Non-sterile | |
0.025 to 0.34 mL | |
20 | |
Black |
With Lid | |
0.34 cm2 | |
Black, Clear Bottom | |
96 | |
Poly-L-Lysine | |
Flat Bottom (F-well) | |
20/Cs. |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.