Learn More
Greiner Bio-One CELLCOAT™ Cell Culture Flasks
Provide enhanced adherence of cells and a cell environment mimicking the in vivo state of the cell. Greiner Bio-One™ CELLCOAT™ Cell Culture Flasks can be used for studies of cellular differentiation, cell-matrix interactions, receptor-ligand binding, gene expression and regulation, embryogenesis and organ development, drug screening and sensitivity assays and culture of primary tumors.
£813.00
Specifications
Material Polystyrene
Neck Style Canted
Surface Coating Collagen Type I Protein
Graduated Yes
View More Specs
Products 1
Product Code Brand Capacity (Metric) Includes Volume (Metric) Working Price Quantity & Availability  
Product Code Brand Capacity (Metric) Includes Volume (Metric) Working Price Quantity & Availability  
11719492
View Documents
Greiner Bio-One
50 mL Red filter screw cap 5 to 10 mL
£813.00
Pack of 50
Description
Description
  • Promotion of cell adhesion, proliferation and growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
  • Shelf life: 24 months at room temperature
  • Cell cultivation in serum-free or serum-reduced medium
  • Rat tail Collagen Type I
  • Increase in isolation and cultivation efficiency
  • Ready-to-use products: immediate use, time-saving 
  • Consistent quality 
  • Storable at room temperature
Specifications
Specifications
Polystyrene
Canted
Collagen Type I Protein
Yes