missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CellCoat™ 96-Well, Collagen Type I-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Brand: Greiner Bio-One 655956
Additional Details : Weight : 0.27000kg
Description
- Cell cultivation in serum-free or serum-reduced medium
- Rat tail Collagen Type I
- Increase in isolation and cultivation efficiency
- Ready-to-use products: immediate use, time-saving -
- Consistent quality
- Storable at room temperature
- Growth area per well: 0.34 cm²
- Max. volume: 0.392 ml
- Working volume: 0.025 ml - 0.34 ml
- With lid
- Shelf life: 24 months at room temperature
Specifications
Alphanumeric | |
With Lid | |
Black, Clear Bottom | |
96 | |
Collagen Type I | |
20/Cs. | |
Chimney |
Standard | |
8 x 12 | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) | |
Clear |