Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Cell Culture Multiwell Plates
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Cell Culture Multiwell Plates helps in promotion of cell adhesion and proliferation.
Brand: Greiner Bio-One 662930
Additional Details : Weight : 0.80000kg
Description
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.- Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Clear | |
Polystyrene | |
3.3 mL | |
1.9 sq. cm | |
Rectangular | |
Poly-D-Lysine | |
24 | |
50 | |
85.48 mm |
Closure Included | |
Flat | |
Clear | |
Polystyrene | |
Non-sterile | |
Multi-well Cell Culture Plate | |
Lid | |
127.76 mm | |
19 mm |