Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks
GREENER_CHOICE
Product Code. 11799482
Click to view available options
Culture Area:
175 cm2
25 cm2
Volume (Metric) Working:
50 mL
650mL
Unit Size:
Pack of 40
Pack of 50
Product Code. 11799482
Brand: Greiner Bio-One 661940
Only null left
Add to basket
Add to basket
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks helps in promotion of cell adhesion and proliferation
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
  • Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
  • For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
  • Ready-to-use products
  • Includes lid with condensation rings
TRUSTED_SUSTAINABILITY
Specifications
Surface Coating Poly-D-Lysine
Closure Type Plug Seal Cap
Culture Area 175 cm2
Volume (Metric) Working 650mL