Learn More
Corning™ CellCube™ Modules, With Aseptic Connectors
Provides a fast, simple, and compact method for the mass culture of attachment-dependent cells.
Brand:  Corning™ 3232
Additional Details : Weight : 4.27500kg
 View more versions of this product
Product Code. 17209109
  • £718.66 / Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Uses a tissue culture-treated growth surface for cell attachment, and continually perfuses the cells with fresh medium for increased cell productivity
  • The CellCube system provides an environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen with low differential gradients across all cells within the modules.
  • The CellCube modules provide a traditional tissue culture-treated surface or Corning CellBIND™ surface for the growth of attachment dependent cells.
Specifications
Specifications
Growth Surface and Aseptic Connectors
1/Pk.
21,250 cm²
1/Cs.
Sterile
25-Layer