missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellCube Module

Designed to enhance cell attachment under difficult conditions

Brand:  Corning™ 3321

Code : 67

Additional Details : Weight : 20.41164kg

Product Code. 10693143

  • £4080.00 / Pack of 2
In Stock
to see stock.
Add to basket

Description

Description

Designed to enhance cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yield. Standard 33mm threaded caps have 0.2μm non-wettable membrane sealed in to allow gas exchange. 26mm diameter filling parts allow direct access to the chamber.
  • CellBIND™ surface treatment
  • Enhanced cell attachment
  • Certified non-pyrogenic and sterilized by gamma radiation
Specifications

Specifications

5000 to 7601 mL
2
Yes
1Pack
Polystyrene
CellBind CellStack
25,440 cm²
Gamma radiation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers