Learn More
Corning™ CellCube™ Systems with Corner Clips
GREENER_CHOICEBIOPHARMA
The CellCube™ module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.
Brand:  Corning™ 3264
 View more versions of this product
Product Code. 17279109
  • £2330.00 / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
CellCube™ modules provide:
  • a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
  • 85,000 cm2 polystyrene growth surface optimally treated with the Corning™ CellBIND™ surface for the growth of attachment dependent cells.
  • a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
  • an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
Corning™ CellBIND™ Surface Benefits:
  • Improves cell attachment leading to increased cell growth and yields
  • Results in more consistent and even cell attachment
  • Helps adapt cells to reduced serum or serum-free conditions
  • Reduces premature cell detachment from confluent cultures
  • May eliminate the need for tedious, time-consuming, expensive and low stability biological coatings
  • Not a coating, requires no refrigeration or special handling and is stable at room temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 L
Square
Sterile
Tissue Culture-treated
Corner clips
No
85,000 cm2
100
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ CellCube™ Systems with Corner Clips > CellCube; 100-layers, Tissue Culture Treated, with Corner Clips, 6 L