missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CellEvent™ Caspase-3/7 Green Flow Cytometry Assay Kit

The CellEvent™ Caspase-3/7 Green Flow Cytometry Assay Kit enables the flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells.

Brand:  Molecular Probes™ C10740

95.95 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 15583329

  • £105.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The kit includes the novel fluorogenic substrate CellEvent™ Caspase-3/7 Green Detection Reagent, as well as SYTOX™ AADvanced™ Dead Cell Stain.

• Caspase-3/7 specific—includes the recognition sequence for activated caspase-3 and caspase-7
• Easy identification—clearly identify live, dead, and apoptotic cell populations
• Quick analysis—washing and fixation is not required
• Multicolor compatibility—combine with other dyes excitable by the 488 nm laser or other lasers

CellEvent™ Caspase-3/7 Green Detection Reagent is a cell-permeant reagent that consists of a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. During apoptosis, caspase-3 and caspase-7 proteins are activated and able to cleave the caspase 3/7 recognition sequence encoded in the DEVD peptide. Cleavage of the recognition sequence and binding of DNA by the reagent labels the apoptotic cells with a bright, fluorogenic signal that has absorption/emission maxima of ∼511/533 nm. When used together with the SYTOX™ AADvanced™ Dead Cell Stain, apoptotic cells can be easily discriminated from live and necrotic cells.

Because no single parameter defines apoptosis in all systems, we strongly suggest using a combination of different measurements for reliable detection of apoptosis.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

20 Assays
Caspase Assay Kit
488
Wet Ice
Fluorescence
Attune™ Acoustic Focusing Cytometer, Flow Cytometer
Other Label(s) or Dye(s)
Kit
Contains 1 vial CellEvent™ Caspase 3/7 Green Detection Reagent, 1 vial SYTOX AADvanced dead cell stain, and 1 vial of DMSO.

Store all vials in the kit upright, at or below -20deg;C, desiccated, and protected from light.
SYTOX AADvanced: 546/647
CellEvent Caspase 3/7 Green Detection Reagent: 511/533
CellEvent™
Flow Cytometry
CellEvent™ Caspase 3/7 Green Detection Reagent, SYTOX AADvanced dead cell stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.