Learn More
CellFIX™ Tissue Processing Reagent, BD
Used for the fixation of human peripheral blood cell suspensions. BD Biosciences™ CellFIX™ is intended for the fixation such suspensions after immunofluorescence staining with monoclonal antibodies and prior to flow cytometric analysis.
Brand: BD Biosciences 340181
Code : Z1
Additional Details : CAS Number : 50-00-0 Weight : 0.16200kg
Description
- When stored at 20° to 25°C, the reagent concentrate is stable until the expiration date shown on the label.
- Do not use after the expiration date.
- Alteration in the appearance of the reagent, such as precipitation or discoloration, indicates instability or deterioration
Specifications
Cellfix | |
Fixation of human peripheral blood cell suspensions after immunofluorescence staining with monoclonal antibodies and prior to flow cytometric analysis | |
50 mL |
Safety and Handling
- BD CellFIX
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : formaldehyd e...%, sodium azide