missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ DMEM/Ham's F-12 Mix
BIOPHARMA

Product Code. 15323561
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
Glutamine
Glutamine, HEPES
Without Additives:
No Phenol Red
None
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15323561

Brand: Corning™ 15090CV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Corning™ cellgro™ DMEM/Ham's F-12 Mix support cell culture growth and substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Original formulation was used to culture embryonic mouse cells
  • Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
  • Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
  • Tested to determine pH and osmolality
  • Low levels of endotoxin, and absence of contamination
  • Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Mycoplasma: Tested Negative
  • Sterility: Pass
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM and Ham's F-12, 50/50 Mix
With Additives Glutamine
Without Additives None
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 500 mL
Endotoxin Level <0.25EU/mL
Formulation L-Glutamine, Sodium Pyruvate, Phenol Red, Sodium Bicarbonate
Content And Storage 2 to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.