missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DMEM (Dulbecco's Modified Eagle's Medium)
BIOPHARMA

Product Code. 15303621
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
1 x 10 L
1 x 20 L
With Additives:
Glucose
Glucose, Glutamine
Glucose, Glutamine, Sodium Pyruvate
Glucose, Sodium Pyruvate
Glucose, Sodium Pyruvate, HEPES
Glutamine, Glucose
Without Additives:
No Glutamine
No Glutamine, No Phenol Red
No Glutamine, No Sodium Pyruvate
No Sodium Pyruvate
None
Product Type:
DMEM Media, 1 g/L Glucose
DMEM Media, 4.5 g/L Glucose
Unit Size:
10 L
20 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15303621

Brand: Corning™ 15013LB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Corning™ DMEM (Dulbecco's Modified Eagle's Medium) support cell culture growth and substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Original formulation was used to culture embryonic mouse cells
  • Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
  • Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
  • Tested to determine pH and osmolality
  • Low levels of endotoxin, and absence of contamination
  • Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
  • Osmolality: 335±mOsm/kg H2O
  • Mycoplasma: Tested Negative
  • Sterility: Pass
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM Media, 4.5 g/L Glucose
With Additives Glucose, Sodium Pyruvate
Without Additives No Glutamine
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 1 x 20 L
Endotoxin Level <0.25EU/mL
Formulation Inorganic Salts: 200mg/L CaCl2 (anhydrous), 0.1mg/L Fe(NO3)3.9H2O, 400mg/L KCl, 97.7mg/L MgSO4 (anhydrous), 6400mg/L NaCl, 125mg/L NaH2PO4.H2O, 3700mg/L NaHCO3; Amino Acids: 84mg/L L-Arginine. HCl, 62.57mg/L L-Cystine.2HCl, 30mg/L Glycine, 42mg/L L-Histidine.HCl.H2O, 104.8mg/L L-Isoleucine, 104.8mg/L L-Leucine, 146.2mg/L L-Lysine. HCl, 30mg/L L-Methionine, 66mg/L L-Phenylalanine, 42mg/L L-Serine, 95.2mg/L L-Threonine, 16mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 20mg/L L-Valine; Vitamins: 4mg/L D-Calcium pantothenate, 4mg/L Choline chloride, 4mg/L Folic Acid, 7.2mg/L i-Inositol, 4mg/L Nicotinamide, 4mg/L Pyridoxine. HCl, 0.4mg/L Riboflavin, 4mg/L Thiamine. HCl; Other: 4500mg/L D-Glucose, 15mg/L Phenol red. Na, 110mg/L Sodium pyruvate; Added: 584mg/L L-glutamine Powder, 20 mL/L 200 mM Solution.
Content And Storage 2 to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.