Learn More
Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform
Fast, automated cellular imaging and analysis platform designed for quantitative microscopy.
Brand: Thermo Scientific™ CX7A1110
Alert:
For research use only
Description
Integrated system for high content analysis
The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.
Features of the CellInsight CX7 HCA Platform
- 7-channel fluorescent imaging
- 5-channel bright-field imaging
- 7-channel confocal imaging
- Software and laser-based autofocus for consistent scan times
- Configured for fully automated plate handling and scanning
Capabilities
- Addresses hundreds of assay types with flexible software and hardware tools
- Scan and analyze microscope slides or most SBS-standard format microplates
- Use HCS Studio software for integrated instrument control and data analysis
- Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
- Compatible with Molecular Probes™ fluorescent detection reagents
- Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
- Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
- Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
- Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
- HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
- Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler
Specifications
CellInsight | |
High Content Instrument | |
Chromogenic, Fluorescence | |
• 1 Reader | |
Room Temperature |
CellInsight™ CX7 High Content Analysis Platform | |
Assorted Colors | |
Microplate, Slide(s) | |
Less hazardous, less waste and use of fewer resources |