missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CellInsight™ CX7 LZR Pro High Content Screening Platform
The Thermo Scientific CellInsight CX7 LZR Pro High Content Screening (HCS) Platform is a fast, laser light engine-based, automated cellular imaging and analysis platform designed for life scientists who require on-the-fly quantitative analysis.
Brand: Thermo Scientific™ HCSDCX7LZRPRO
This item is not returnable.
View return policy
Description
- A fast, laser light engine-based, automated cellular imaging and analysis platform designed for life scientists who require on-the-fly quantitative analysis
- The CellInsight CX7 LZR Pro Platform offers a more powerful and integrated way to develop and automate high content assays using a combination of 5-color bright-field and 7-color fluorescence imaging in confocal or wide-field modes
- With laser illumination, the CellInsight LZR Pro Platform enables researchers to image faster in both wide-field and confocal mode
Features of the CellInsight CX7 LZR Pro Platform:
- Back-illuminated sCMOS camera offers a 95% peak quantum efficiency and -20°C cooling
- New validated applications including Cell Painting and FirePlex-HT immunoassay
- Olympus X-line objective imaging to improve imaging quality and assay performance
- EurekaScan Finder double-pass functionality for on-the-fly seek and find of spheroids, tissue, and rare cellular events
- HCS Studio 5.0 with over 40 life science bio-applications and real-time analysis for fastest time-to-data possible.
- 7-channel fluorescent imaging with laser illumination
- 5-channel bright-field imaging
- 7-channel confocal imaging with laser illumination
- Software and laser-based autofocus for consistent scan times
- Configured to accept optional modules for fully automated plate handling and live cell on-board environmental control
Capabilities
- Use integrated HCS Studio software for instrument control and data analysis
- Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
- Addresses hundreds of assay types with flexible software and hardware tools
- Scan and analyze microscope slides or most SBS-standard format microplates, including 6-well through 1536-well formats
- Compatible with Molecular Probes fluorescent detection reagents
Hardware and software
- High-output laser excitation solid state light
- Broad reagent options with 7-color excitation from violet to near IR for assay multiplexing
- Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
- Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
- HCS Studio 5.0 Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
- Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler (not included)

Specifications
CellInsight | |
Highly Sensitive Monochrome CMOS | |
0 to 3000 VDC, 0 to 250 VAC |
Linear, Orbital, Double Orbital | |
1 each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction