missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CellMask™ Green Actin Tracking Stain
Stain and track intracellular F-actin with CellMask Green Actin Tracking Stain in live or fixed cells ICC, IHC, and 3D cell culture. The stain easily permeates live cell membranes, is retained within the cells after loading, and shows no detectable effects on viability after 24 hours of incubation.
£279.00 - £606.00
Specifications
Color | Green |
---|---|
Excitation Wavelength Range | 503 nm |
Dye Type | FITC/GFP |
For Use With (Equipment) | Fluorescence Microscope, High Content Analyzer |
Detection Method | Fluorescence Microscopy |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17163269
|
Invitrogen
A57243 |
1 x 60 Assays |
£279.00
Pack of 60 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17183269
|
Invitrogen
A57246 |
5 x 60 Assays |
£606.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Stain and track intracellular F-actin with CellMask Green Actin Tracking Stain in live or fixed cells in immunofluorescence (IF), immunocytochemistry (ICC), and immunohistochemistry (IHC) protocols, and in 3D cell culture. The CellMask Green Actin Tracking Stain fluorescently labels polymerized/filamentous actin (F-actin) in live or fixed cells because it is designed to readily permeate live cells, resulting in bright and uniform labeling. CellMask Green Actin Tracking Stain can be used to track F-actin in live cells for 24 hours or more. Live cells stained with CellMask Green Actin Tracking Stain can be fixed; the stain can also be used on already fixed cells. Cell Mask Green Actin Tracking Stain can be multiplexed with antibodies in IF, ICC, and IHC protocols, and can also be used in 3D cell culture for 3D imaging.Features of Cell Mask Green Actin Tracking Stain include:
• Stains F-actin in live or fixed cells for fluorescence 2D monolayer or 3D imaging experiments
• Can be multiplexed with antibodies in IF/ICC/IHC experiments
• Fluorescence maxima (Excitation/Emission): 503/512 nm; can be detected with FITC/GFP filter
CellMask actin tracking stains are available in three colors: green (Cat. No. A57243, A57246) detected with a FITC/GFP filter (Ex 503 nm/Em 512 nm), orange (Cat. No. A57244, A57247), detected with a TRITC/RFP filter (Ex 545 nm/Em 570 nm), and deep red (Cat. No. A57245, A57248) detected with a CY5/Deep Red filter (Ex 652 nm/Em 669 nm).
CellMask actin tracking stains detect F-actin with a targeting structure that closely resembles jasplakinolide. They do not label globular actin (G-actin) or monomer actin (M-actin). CellMask actin tracking stains easily pass through live cell membranes and are retained within live cells after loading. As demonstrated in the figures below, incubation with CellMask actin tracking stains has no detectable effect on viability of various cell lines after 24 hours of incubation.
CellMask actin tracking stains detect F-actin in live cells or tissue, which can then be fixed. In addition, CellMask actin tracking stains can be used to detect F-Actin in paraformaldehyde fixed cells or tissue (freshly fixed or cryo frozen only) and can be multiplexed with antibody detection. CellMask actin tracking stains can also be used in 3D cell culture for 3D imaging.
Specifications
Green | |
FITC/GFP | |
Fluorescence Microscopy | |
512 nm | |
Room Temperature | |
Fluorescent Dye | |
Actin, Cytoskeleton |
503 nm | |
Fluorescence Microscope, High Content Analyzer | |
CellMask™ Green Actin Tracking Stain | |
CellMask™ | |
F-Actin | |
Actin |
For Research Use Only. Not for use in diagnostic procedures.