Learn More
Greiner Bio-One CELLMASTER Polyethylene Terephthalate (PET) Roller Bottles - Short Form
For use in cell culture. Greiner Bio-One CELLMASTER Polyethylene Terephthalate (PET) Roller Bottles - Short Form is suitable for many adherent cell lines, is stable and gas permeable.
£187.00
Products 1
Product Code Brand Diameter (Metric) No. per Case Price Quantity & Availability  
Product Code Brand Diameter (Metric) No. per Case Price Quantity & Availability  
10481292
View Documents
Greiner Bio-One
680180
122 mm 1
£187.00
Pack of 18
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co