Learn More
Nexcelom™ Cellometer Disposable Counting Chambers (PD100 Slides)
The fixed 20 µL sample size allows for simple, automated calculation of cell concentration following imaging and counting port
Brand: Nexcelom™ CHT4-003
Code : Z1
Additional Details : Weight : 0.10000kg
Description
- Each all-plastic, disposable slide contains two sample counting chambers with precisely controlled height
- After loading the sample, the slide is inserted into a Cellometer System
- Cells are imaged directly from the center of the counting chamber
- Image-based counting with disposable counting chambers offers several key advantages:
- No potential clogging
- Ideal for fragile samples, such as hepatocytes
- No washing
- No potential cross-contamination
Specifications
20 μL | |
10 individual boxes | |
500 |