Learn More
BINDER™ CELLROLL Set
Product Code. 17313683 Shop All BINDER Products
Click to view available options
Model:
CBF 170
CBF 260
Quantity:
4 Roller Bottles/Pk.
6 Roller Bottles/Pk.
Unit Size:
Each
Product Code. 17313683
Brand: BINDER™ 86120023
Only null left
Add to basket
Add to basket
CELLROLL Set is a modular, expandable roller bottle system for cell cultivation.
  • Adjustable direction of rotation, speed of rotation, pause times, and interval cycles
  • All drive rollers for the bottles run without vibrations on special ball bearings
  • Driven by a stepper motor with a soft start and stop phase
  • Control unit is outside the incubator and is connected to the interior motor unit via an electric access port
Specifications
Description CELLROLL Set - modular, expandable roller bottle system for cell cultivation, 4 roller bottles
For Use With (Equipment) BINDER Series CB and CBF
Material Stainless Steel
Type Cell Roller
For Use With (Application) Cell Cultivation
Model CBF 170
Quantity 4 Roller Bottles/Pk.