Learn More
Molecular Probes™ CellROX™ Green Flow Cytometry Assay Kit
CellROX™ Green Flow Cytometry Assay Kit
Brand:  Molecular Probes™ C10492
354.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 15258236
  • £370.00 / 1 set
Explore more special offers
Description
Description
The CellROX Green Flow Cytometry Assay Kit enables the flow cytometric detection of reactive oxygen species (ROS) in live cells. The kit includes the novel, fluorogenic CellROX Green Reagent, as well as SYTOX Red Dead Cell Stain, N-acetyl cysteine (an antioxidant, for negative control), and tert-butyl hydroperoxide solution (TBHP, an inducer of ROS).
The CellROX Green Flow Cytometry Assay Kit features:
• Fluorogenic probe formulated for flow cytometry that is oxidized in the presence of ROS
• Multicolor compatibility—minimal overlap with fluorophores excited by other laser lines, allowing easy multiplexing with other reagents
• Simple protocol—cells can be stained in complete media or other appropriate buffer, no need for serum free media
The CellROX Green Detection Reagent is cell-permeable and non-fluorescent or very weakly fluorescent while in the reduced state. Upon oxidation, the reagent binds DNA and exhibits a strong fluorogenic signal that has absorption/emission maxima of 508/525 nm and remains localized to the nucleus and mitochondria. When used together with the included SYTOX Red Dead Cell Stain, oxidatively stressed and nonstressed cells are reliably distinguished from dead cells by flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube
100 assays
CellROX green: 508/525, SYTOX red: 640/658