Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Product Code. 10450142
Click to view available options
Color:
Clear
Unit Size:
Pack of 40
Product Code. 10450142
Brand: Greiner Bio-One 781165
Only null left
Add to basket
Add to basket
Color:Clear; Sterility:Sterile; Lid:Without Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-cytotoxicCompatible with automated systemsFlexible working volume
  • Rounded square well profile
  • Stackable
  • Alphanumeric well cod
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
No. per Case 40
No. per Pack 10