missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks
GREENER_CHOICE

Product Code. 10716172
Click to view available options
Capacity (Metric):
50 mL
250 mL
650 mL
Closure Type:
Screw
White Filter Screw Cap
Unit Size:
Pack of 120
Pack of 200
Pack of 40
This item is not returnable. View return policy

Product Code. 10716172

Brand: Greiner Bio-One 658195

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to culture cell suspended in a growth medium. Greiner Bio-One™ CELLSTAR Suspension Cell Culture Flasks are manufactured from polystyrene for optimum visibility and strength.

  • Hydrophobic surface, ideal for suspension cultures and hybridoma
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Graduation on the side
  • Canted neck
  • Easily distinguished by their white screw caps
  • White standard screw cap with ventilation position
  • TC surface treatment: -/suspension
  • Sterile and user friendly packaging
TRUSTED_SUSTAINABILITY

Specifications

Closure Color White
Capacity (Metric) 250 mL
Material Polystyrene
No. per Case 120 Pack
No. per Pack 120
Graduated Yes
Closure Type White Filter Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.