missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellTrace™ Far Red Cell Proliferation Kit, for flow cytometry

CellTrace™ Far Red Cell Proliferation Kit, for flow cytometry

Brand:  Invitrogen™ C34564

391.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15396613

  • £441.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

CellTrace™ Far Red Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry.

  • Superior performance—bright, single-peak staining enables visualization of multiple generations
  • Long-term signal stability—well-retained in cells for several days post stain
  • Versatile—multiple colors available to easily combine with antibodies or markers of cell function, such as GFP
  • Simple, robust staining protocol

    View a selection guide for all CellTrace™ Cell Proliferation Kits for flow cytometry.

    Superior fluorescent staining
    Successful proliferation analysis by dye dilution requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence after several cell divisions. The intense fluorescent staining provided by CellTrace™ Far Red dye enables the visualization of six or more generations of proliferating cells before the signal is overwhelmed by intrinsic cellular auto-fluorescence. This kit enables consistent, homogeneous staining results with very little fluorescence variation between cells in a population, so distinct generations can be seen without any requirement for complex modeling software.

    Long-term signal retention
    Unlike stains that label the lipid membrane of cells, CellTrace™ Far Red stain easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye offers a consistent signal, even after several days in a cell culture environment. CellTrace™ Far Red dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells.

    Easy multiplexing with other fluorophores
    The red excitation at 630 nm and emission at 661 nm of CellTrace™ Far Red dye make it ideal for multiplexing due to the limited spectral overlap with other common dyes (Alexa Fluor™ 488, FITC, and RPE) and fluorescent proteins (Green Fluorescent Protein (GFP) and mCherry).

    Simple, robust staining protocol
    The CellTrace™ Far Red Cell Proliferation Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1μL of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Continas 9 vials of CellTrace™ Far Red (lyophilized powder) and 1 vial of DMSO (500 μL). Store in freezer (-5 to -30°C).
    Proliferation Assay
    Cell Proliferation Kit
    633/635
    Tube(s)
    180 assays kit
    Fluorescence
    Flow Cytometer
    Contains 9 vials of CellTrace™ Far Red (lyophilized powder) and 1 vial of DMSO (500μL)
    Lyophilized
    CellTrace™
    Room Temperature
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.