missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Cellulose Dialysis Tubing Roll

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.

Brand:  Biodesign™ D002

 View more versions of this product

Product Code. 12787466

  • £292.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.

  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 30.5m rolls
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications

Specifications

Dialysis Tubing Roll
Cellulose
30.5 m
Transparent
For life sciences research
14 kDa
15.5 mm
1.91 mL/cm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers