missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Cellulose Nitrate Membrane Filter
Used for routine microbiological quality control
Brand: Sartorius™ 13006--47------N
98.10 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Used for routine microbiological quality control
- Contains mixture of cellulose nitrate and cellulose acetate
- Assures effective retention
- High flow rates
- Optimal colony growth
- Contrast to colonies for easy and reliable quantification, identification
- Outstanding recovery rates for microorganisms Gamma irradiated at 25kGray
Specifications
Gray, White | |
Cellulose Nitrate (Cellulose Mixed Ester), Cellulose Acetate or Regenerated Cellulose | |
Sterile | |
Hydrophilic | |
47 mm |
Type 130 | |
0.45 μm | |
Individually Packed | |
100/Pk. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MEMBRANE DS.NITRATE 0.45UM47MM FP=100 3.1X 3.1MM
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose