missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11302, 3μm
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11302, 3μm are recommended for a broad range of applications including liquid filtering for sample preparation.
£133.62 - £196.00
Specifications
Color | White |
---|---|
Material | Cellulose Nitrate, Mixed Cellulose Ester |
Pore Size | 3 μm |
Sterility | Non-sterile |
Grade | 11302 |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15952450
|
Sartorius Stedim Biotech™
11302--68------G |
68 mm | |||||||
15962450
|
Sartorius Stedim Biotech™
11302--90------G |
90 mm |
£196.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Cellulose nitrate (CN), mixed cellulose ester (MCE), membrane filters are composed of cellulose nitrate and a small content of cellulose acetate.
- Recommended for a broad range of applications such as the filtration of aqueous solutions or sample preparation.
- Also suitable for air/gas filtration, clarification, particle analysis
- Sterilizable by autoclave, gamma irradiation, or ETO
- Non-sterile, bulk-packed
Specifications
White | |
3 μm | |
11302 | |
25/pk. |
Cellulose Nitrate, Mixed Cellulose Ester | |
Non-sterile | |
Hydrophilic | |
130 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X25 Cellulose Nitrate (Mixed Cellulose Ester) Memb rane Filters / Type 11302, 3 µm, 68 mm
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose