missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ Science Precision Wipes
GREENER_CHOICE

Low-lint, high absorbency and chemically inert wipes for delicate tasks and instrumentation in a range of applications. The 1-ply white-colored Kimtech™ Science Precision Wipes are provided in a convenient pop-up box and are anti-static.

£12.44 - £278.00

Specifications

Product Type Critical Task Wipe
Material Cellulose
Color White
For Use With (Application) Light Duty, Precision Cleaning of Delicate Glassware, Instruments and Surface/Parts
Ply 1-ply
View More Specs

Products 3
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
11778188
GREENER_CHOICE
View Documents
Kimberly-Clark™
7552
20.8 x 11.2 cm (8.2 x 4.4 in.) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10623111
GREENER_CHOICE
View Documents
Kimberly-Clark™
7552
20.8 x 11.2 cm (8.2 x 4.4 in.) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11768188
GREENER_CHOICE
View Documents
Kimberly-Clark™
7551
31 x 28.5 cm (12.2 x 11.2 in.)
£278.00
Pack of 15
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • For delicate equipment, surfaces, optics and materials, our low-lint and high absorbency white wipes have you covered
  • Light duty, white-colored wipes made from 100% pure fiber
  • Provided in a hygienic, user-friendly pop-up box
  • Low-lint absorbency for delicate applications and surfaces
  • Suitable for use in ISO Class 4 or higher environments
  • Suitable for applications in healthcare, manufacturing, optics and electronics
  • Chemically inert and anti-static
Specifications

Specifications

Critical Task Wipe
White
1-ply
Cellulose
Light Duty, Precision Cleaning of Delicate Glassware, Instruments and Surface/Parts
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Kimtech™ Science Precision Wipes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.