Learn More
3M™ Center Adapter Assembly 6864/37000
3M™ Center Adapter Assembly 6864/37000 includes center port adaptor, exhalation valve and lens gasket for the 3M™ Full Face Mask 6000 Series.
Brand: 3M™ 7000002178
17.05 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.21050kg
Description
- Replacement full face respirator center adapter assembly includes center port adaptor, exhalation valve and lens gasket
- Replace damaged parts to ensure correct levels of protection and comfort
- For use with the 3M™ 6000 Series Full Face Respirator
Specifications
Gray | |
Center Adapter Assembly | |
5/Box |