missing translation for 'onlineSavingsMsg'
Learn More

Merck Centrifree Ultrafiltration Device with Ultracel™ PL membrane

Separate free from bound microsolutes. Centrifree™ Ultrafiltration Device is licensed for research use and in vitro diagnostic use with serum, plasma & other biological samples. Features the Ultracel™ cellulose membrane

Brand:  Merck 4104

Product Code. 10238110

  • £838.00 / Pack of 50
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Separate free from bound microsolutes. Centrifree™ Ultrafiltration Device is licensed for research use and in vitro diagnostic use with serum, plasma & other biological samples. Features the Ultracel™ cellulose membrane

The Centrifree™ Ultrafiltration device with Ultracel™ regenerated cellulose membrane is ideal for separating free from bound microsolutes in serum, plasma, and other biological samples. The device is licensed for research use and in vitro diagnostic use. The device holds a sample volume of 1 mL with a hold-up volume of 10 μL.

Features & Benefits:

  • Separate unbound (free) therapeutic drugs, testosterone, thyroxin, etc.
  • Enables accuracy in binding studies
  • Critical for new drug investigations
  • Facilitates deproteinization to reduce complexity or matrix interference of biological samples
  • For research and in vitro diagnostic use
  • Applications:Determination of Free Drugs, Testosterone, Thyroxin or Other Bioactives, Binding Studies, New Drug Investigations

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Clear
    Regenerated Cellulose
    Non-sterile
    Separate free from bound microsolutes
    Ultracel™
    Centrifugal Filters
    95 mm
    Single-use
    30 kDa
    0.92 cm2
    Regenerated Cellulose (RC)
    50Pack
    1 mL
    16 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers