Learn More
OHAUS™ Centrifuge Adapter
Adapter accessory for Centrifuge Multi models
Brand:  OHAUS™ 30559412
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 16632784
  • £206.00 / Pack of 2
In Stock
to see stock.
Description
Description
  • Adapter for various Centrifuge Multi models
  • Various capacities available
Specifications
Specifications
Adapter
Centrifuge Multi 230 V FC5816, Centrifuge Multi 230 V FC5714, Centrifuge Multi 120 V FC5816R, Centrifuge Multi 230 V FC5816R, Centrifuge Multi 120 V FC5816, Centrifuge Multi FC5916R 120 V, and Centrifuge Multi FC5916 120 V
MTB x2
15 x 1.5/2.0 mL
Depth 11 mm