Learn More
OHAUS™ Centrifuge Adapter
Adapter accessory for Centrifuge Multi models
Brand:  OHAUS™ 30559413
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 16642784
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Adapter for various Centrifuge Multi models
  • Various capacities available
Specifications
Specifications
Adapter
Centrifuge Multi 230 V FC5816, Centrifuge Multi 230 V FC5714, Centrifuge Multi 120 V FC5816R, Centrifuge Multi 230 V FC5816R, Centrifuge Multi 120 V FC5816, Centrifuge Multi FC5916R 120 V, and Centrifuge Multi FC5916 120 V
FA x3
250 mL
Depth 60 mm