Learn More
OHAUS™ Centrifuge Adapter
Adapter accessory for Centrifuge Multi models
Brand:  OHAUS™ 30559414
 View more versions of this product
Product Code. 16652784
  • £197.00 / Pack of 2
Explore more special offers
Description
Description
  • Adapter for various Centrifuge Multi models
  • Various capacities available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
Centrifuge Multi 230 V FC5816, Centrifuge Multi 230 V FC5714, Centrifuge Multi 120 V FC5816R, Centrifuge Multi 230 V FC5816R, Centrifuge Multi 120 V FC5816, Centrifuge Multi FC5916R 120 V, and Centrifuge Multi FC5916 120 V
FA x2
175/225 mL
Depth 61 mm