missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Centrifuge Adapter

Adapter accessory for Centrifuge Multi models

Brand:  OHAUS™ 30559416

Additional Details : Weight : 0.36000kg

 View more versions of this product

Product Code. 16662784

  • £166.00 / Pack of 2
In Stock
to see stock.

Description

Description

  • Adapter for various Centrifuge Multi models
  • Various capacities available
Specifications

Specifications

Adapter
Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V
FA x2
250 mL
Depth 60.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers