Learn More
Eppendorf™ Centrifuge Adapters For Rotors for 5810 and 5810R
Available in a variety of configurations to suit a wide range of applications
Brand:  Eppendorf™ 5810770009
 View more versions of this product
Product Code. 10326802
  • £185.74 / Pack of 2
Explore more special offers
Description
Description
  • Adaptors for Eppendorf™ Centrifuges 5810 and 5810R.
  • Compatible with conical or flat tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
180 to 250 mL
62 mm
2 Pack
Flat
Rotors 5810 and 5810R Centrifuge
4