missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Replacement 6 x 1.5mL Rotor

For standard microtubes and adapters. Fisherbrand™ Replacement 6 x 1.5mL Rotors are for use with the Fisherbrand Mini Centrifuge.

Brand:  Fisherbrand™ C1300-RT

Product Code. 11441967

  • £64.45 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Prolongs the life of your centrifuge
  • Manufactured from durable materials
  • Accelerates quickly to 6,000 RPM
Specifications

Specifications

Rotor
6,000 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.