Learn More
Unity Lab Services™ Centrifuge Standard Requalification Service
Click to view available options
For Use With (Equipment):
Centrifuge
Unit Size:
1 unit
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
- Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
- Key tests Min/Max speed, Brief /Extended time, temperature for refrigerated models.
- Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
Specifications
Specifications
| For Use With (Equipment) | Centrifuge |
| Product Type | Requalification Service |