missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ CentriVap -50 Cold Trap with Acrylic Lid
The CentriVap Cold Trap collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump and the laboratory environment from the corrosive effects of vapors and fumes emitted from samples.
Brand: Labconco™ 7811035
This item is not returnable.
View return policy
Description
The CentriVap -50°C Cold Trap collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump and the laboratory environment from the corrosive effects of vapors and fumes emitted from samples.Specifications
CE Marked | |
Aqueos Sample Applications | |
-40°C | |
14.5 in. | |
61.4 cm | |
35.0 cm | |
0 to 300 V |
Stainless Steel | |
British (UK) | |
14.5 x 24.2 x 13.8 in. (36.8 x 61.4 x 35.0 cm) | |
24.2 in. | |
13.8 in. | |
230 V 50 Hz 2.4 amp (International) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction