missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Centrivap Ultra Low Cold Trap
The CentriVap -84°C Cold Trap collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump and the laboratory environment from the corrosive effects of vapors and fumes emitted from samples.
Brand: Labconco™ 7460035
This item is not returnable.
View return policy
Description
The CentriVap -84°C Cold Trap collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump and the laboratory environment from the corrosive effects of vapors and fumes emitted from samples.Specifications
CE Marked | |
Low Freezing Point Solvents | |
-40°C | |
14.5 x 22.0 x 20.9 in. (36.8 x 55.9 x 53.1 cm) | |
36.8 cm | |
55.9 cm | |
53.1 cm | |
0 to 300 V |
Stainless Steel | |
British (UK) | |
4 L | |
14.5 in. | |
22.0 in. | |
20.9 in. | |
230 V 50 Hz 6 amp (International) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction