missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ ceramus™ HF Dispenser
Specially developed for dispensing hydrofluoric acid and other aggressive media.
Marque: Hirschmann™ 9342300
Code : 03
Informations supplémentaires : Weight : 0.47250kg
Description
- Volume can be variably adjusted from 2 to 10mL
- Piston, cylinder, valve seats and balls are made of high-purity aluminum oxide ceramic
- Valve springs made of platinum-iridium
- Certified conformity
- With reagent circulation system
- Suitable for steam sterilization up to 121°C
- 45 screw thread
- Provided with 3 threaded adapters (A 32, S 40, A 38), 1 suction hose with union nut, 1 ejection unit and 1 fitting wrench
- With individual serial number and calibration certificate
Spécification
Bottletop Dispenser | |
High Purity Ceramic | |
Dispensing aggressive media | |
3 threaded adapters (A 32, S 40, A 38), 1 suction hose with union nut, 1 ejection unit, 1 fitting wrench, individual serial number, calibration certificate | |
HF |
Calibration certificate, certified conformity | |
2 to 10 mL | |
Bottle | |
0.25 mL | |
0.6% |