missing translation for 'onlineSavingsMsg'
Learn More
Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11321198

Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11321198
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11321198

Brand: Thermo Scientific Alfa Aesar 010141.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Color Silver to Gray
UN Number UN1333
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.42g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Cerium foil, 0.1mm (0.004 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT25K019 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number010141.FF,010141.FI
Lot NumberB27N19 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010141.FF,010141.FI
Lot NumberE01R043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010141.FF,010141.FI
Lot NumberQ23A031 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number010141.FF,010141.FI
Lot NumberR22B036 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number010141.FF,010141.FI
compliance-icons
Product Identifier
  • Cerium foil
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.